4,5-Dimethoxy-2-nitrocinnamic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(4,5-Dimethoxy-2-nitrophenyl)acrylic acid
OR1678-1G
APOSOR1678-1GEA
21.9
EUR
APOSOR1678-1G
4,5-Dimethoxy-2-nitrocinnamic acid 98%
4,5-Dimethoxy-2-nitrocinnamic acid
Formel:
C₁₁H₁₁NO₆ Molecular Weight: 253,21 g/mol Schmelzpunkt: 293 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017017 CAS-Nummer: 20567-38-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...