4-Chloroindole-3-carbaldehyde 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-1H-indole-3-carbaldehyde

OR1683-5G
APOSOR1683-5GEA 126 EUR
APOSOR1683-5G
4-Chloroindole-3-carbaldehyde 98%
4-Chloroindole-3-carbaldehyde
Formel: C₉H₆ClNO
Molecular Weight: 179.6030 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05864704
CAS-Nummer: 876-72-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR