tert-Butyl-4-chloro-3-formyl-1H-indole-1-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-3-formyl-1H-indole, N-BOC protected

OR1684-1G
APOSOR1684-1GEA 119 EUR
APOSOR1684-1G
tert-Butyl-4-chloro-3-formyl-1H-indole-1-carboxylate 98%
tert-Butyl-4-chloro-3-formyl-1H-indole-1-carboxylate
Formel: C₁₄H₁₄ClNO₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 914349-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR