6-Bromo-1H-indole-2-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR16855-500MG
APOSOR16855-500MGEA
342
EUR
APOSOR16855-500MG
6-Bromo-1H-indole-2-carbaldehyde
6-Bromo-1H-indole-2-carbaldehyde
Formel:
C₉H₆BrNO Schmelzpunkt: 181…183 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06738308 CAS-Nummer: 105191-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...