5-Chloro-1-benzothiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chlorobenzo[b]thiophene-2-carboxylic acid

OR16860-1G
APOSOR16860-1GEA 232 EUR
APOSOR16860-1G
5-Chloro-1-benzothiophene-2-carboxylic acid
5-Chloro-1-benzothiophene-2-carboxylic acid
Formel: C₉H₅ClO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01927183
CAS-Nummer: 13771-75-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR