Methyl-4-chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-Chloro-5-azaindole-2-carboxylate

OR16875-1G OR16875-250MG
APOSOR16875-1GEA 133 EUR
APOSOR16875-1G APOSOR16875-250MG
Methyl-4-chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylate
Methyl-4-chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylate
Formel: C₉H₇ClN₂O₂
Molecular Weight: 210.62 g/mol
Schmelzpunkt: 224…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09870065
CAS-Nummer: 688357-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR