3-(2-Methyl-1,3-dioxolan-2-yl)-1-propanamine

Lieferant: Apollo Scientific
Gefahr

OR16878-1G
APOSOR16878-1GEA 538 EUR
APOSOR16878-1G
3-(2-Methyl-1,3-dioxolan-2-yl)-1-propanamine
3-(2-Methyl-1,3-dioxolan-2-yl)-1-propanamine
Formel: C₇H₁₅NO₂
Molecular Weight: 145.2 g/mol
Siedepunkt: 91…92 °C/14mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD09923637
CAS-Nummer: 66442-97-5
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR