tert-Butyl-4-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

OR1688-250MG
APOSOR1688-250MGEA 0 EUR
APOSOR1688-250MG
tert-Butyl-4-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate 98%
tert-Butyl-4-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate
Formel: C₁₄H₁₆BrNO₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 914349-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR