tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

OR1689-250MG
APOSOR1689-250MGEA 0 EUR
APOSOR1689-250MG
tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate 98%
tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate
Formel: C₁₄H₁₆BrNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05864710
CAS-Nummer: 905710-14-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR