2-Amino-5-brombenzaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromoanthranilaldehyde

OR16893-5G
APOSOR16893-5GEA 74.1 EUR
APOSOR16893-5G
2-Amino-5-brombenzaldehyd
2-Amino-5-bromobenzaldehyde
Formel: C₇H₆BrNO
Molecular Weight: 200,00 g/mol
Schmelzpunkt: 75…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10696879
CAS-Nummer: 29124-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR