2-Brom-5-nitrobenzaldehyd 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-nitrobenzenecarbaldehyde

OR16898-5G
APOSOR16898-5GEA 21.9 EUR
APOSOR16898-5G
2-Brom-5-nitrobenzaldehyd 95%
2-Bromo-5-nitrobenzaldehyde
Formel: C₇H₄BrNO₃
Molecular Weight: 230,0200 g/mol
Schmelzpunkt: 103…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00462865
CAS-Nummer: 84459-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR