5-Amino-2-chloropyridine-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-amino-2-chloropyridine-4-carboxylic acid, 5-Amino-2-chloroisonicotinic acid, 3-Amino-6-chloroisonicotinic acid

OR16903-1G
APOSOR16903-1GEA 26.1 EUR
APOSOR16903-1G
5-Amino-2-chloropyridine-4-carboxylic acid
5-Amino-2-chloropyridine-4-carboxylic acid
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172,5700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659499
CAS-Nummer: 58483-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR