6-Amino-5-bromonicotinic acid
Lieferant: Apollo Scientific
OR16907-1G
APOSOR16907-1GEA
43.5
EUR
APOSOR16907-1G
6-Amino-5-bromonicotinic acid
6-Amino-5-bromonicotinic acid
Formel:
C₆H₅BrN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109515 CAS-Nummer: 180340-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...