3-Chloroimidazo[1,2-a]pyridine-6-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR16910-1G
APOSOR16910-1GEA 354 EUR
APOSOR16910-1G
3-Chloroimidazo[1,2-a]pyridine-6-carbohydrazide
3-Chloroimidazo[1,2-a]pyridine-6-carbohydrazide
Formel: C₈H₇ClN₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD11100124
CAS-Nummer: 1211364-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR