Methyl-4-(3-thienyl)benzoate

Lieferant: Apollo Scientific

OR16911-1G
APOSOR16911-1GEA 79.9 EUR
APOSOR16911-1G
Methyl-4-(3-thienyl)benzoate
Methyl-4-(3-thienyl)benzoate
Formel: C₁₂H₁₀O₂S
Molecular Weight: 218,27 g/mol
Schmelzpunkt: 160…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05863312
CAS-Nummer: 20608-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR