6,8-Dibromimidazo[1,2-a]pyrazin

Lieferant: Apollo Scientific
Achtung

OR16939-100G OR16939-25G OR16939-5G
APOSOR16939-100GEA 430 EUR
APOSOR16939-100G APOSOR16939-25G APOSOR16939-5G
6,8-Dibromimidazo[1,2-a]pyrazin
6,8-Dibromoimidazo[1,2-a]pyrazine
Formel: C₆H₃Br₂N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08460056
CAS-Nummer: 63744-22-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR