6-Methoxy-1H-indole-3-carbaldehyde 98%

Lieferant: Apollo Scientific
Achtung

OR1694-5G
APOSOR1694-5GEA 68.3 EUR
APOSOR1694-5G
6-Methoxy-1H-indole-3-carbaldehyde 98%
6-Methoxy-1H-indole-3-carbaldehyde
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Schmelzpunkt: 186…188 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03848058
CAS-Nummer: 70555-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR