4-Amino-1H-indazol

Lieferant: Apollo Scientific

OR16942-1G
APOSOR16942-1GEA 26.1 EUR
APOSOR16942-1G
4-Amino-1H-indazol
4-Amino-1H-indazole
Formel: C₇H₇N₃
MDL: MFCD03305710
CAS-Nummer: 41748-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR