2-Amino-4,5-dichlorophenol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,5-Dichloro-2-hydroxyaniline

OR16946-1G
APOSOR16946-1GEA 94.4 EUR
APOSOR16946-1G
2-Amino-4,5-dichlorophenol
2-Amino-4,5-dichlorophenol
Formel: C₆H₅Cl₂NO
Molecular Weight: 178.016 g/mol
Schmelzpunkt: 170…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12406838
CAS-Nummer: 28443-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR