3-Iodopyrazine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR16959-100MG
APOSOR16959-100MGEA 21.9 EUR
APOSOR16959-100MG
3-Iodopyrazine-2-carboxylic acid
3-Iodopyrazine-2-carboxylic acid
Formel: C5H3IN2O2
Lagertemperatur: Raumtemperatur
MDL: MFCD13193281
CAS-Nummer: 212471-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR