4-(Methylamino)benzonitril

Lieferant: Apollo Scientific
Achtung

OR16971-25G
APOSOR16971-25GEA 128 EUR
APOSOR16971-25G
4-(Methylamino)benzonitril
4-(Methylamino)benzonitril
Formel: C₈H₈N₂
Molecular Weight: 132.16 g/mol
Schmelzpunkt: 87…91 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00016381
CAS-Nummer: 4714-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR