3-Phenylpyrrolidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR16975-1G
APOSOR16975-1GEA
134
EUR
APOSOR16975-1G
3-Phenylpyrrolidine hydrochloride
3-Phenylpyrrolidine hydrochloride
Formel:
C₁₀H₁₄ClN Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179094 CAS-Nummer: 857281-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...