(Iodmethyl)cyclopentan 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR16976-1G
OR16976-250MG
APOSOR16976-1GEA
55.1
EUR
APOSOR16976-1G
APOSOR16976-250MG
(Iodmethyl)cyclopentan 98%
(Iodomethyl)cyclopentane
Formel:
C₆H₁₁I Molecular Weight: 210.06 g/mol Siedepunkt: 156…158 °C Dichte: 1.604 g/cm³ (25 °C) Flammpunkt: 70 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03453190 CAS-Nummer: 27935-87-1 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...