Bis(4-nitrophenyl)carbonat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Bis(4-nitrophenyl) carbonate
OR16983-100G
OR16983-25G
OR16983-500G
APOSOR16983-100GEA
49.3
EUR
APOSOR16983-100G
APOSOR16983-25G
APOSOR16983-500G
Bis(4-nitrophenyl)carbonat 98%
Bis(4-nitrophenyl)carbonat
Formel:
C₁₃H₈N₂O₇ Schmelzpunkt: 136…139 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00007322 CAS-Nummer: 5070-13-3 EINECS: 225-775-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...