6-Aminopyridin-3-ol hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-hydroxypyridine hydrochloride

OR16984-1G
APOSOR16984-1GEA 137 EUR
APOSOR16984-1G
6-Aminopyridin-3-ol hydrochloride 95%
6-Aminopyridin-3-ol hydrochloride
Formel: C₅H₇ClN₂O
Molecular Weight: 146,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11506259
CAS-Nummer: 856965-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR