5-Bromo-3-chloropyridine-2-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-3-chloropicolinic acid
OR16996-1G
OR16996-5G
OR16996-25G
APOSOR16996-1GEA
21.9
EUR
APOSOR16996-1G
APOSOR16996-5G
APOSOR16996-25G
5-Bromo-3-chloropyridine-2-carboxylic acid 97%
5-Bromo-3-chloropyridine-2-carboxylic acid
Formel:
C₆H₃BrClNO₂ Molecular Weight: 236.46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12827552 CAS-Nummer: 1189513-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...