5-Bromo-3-chloropyridine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-chloropicolinic acid

OR16996-1G OR16996-5G OR16996-25G
APOSOR16996-1GEA 21.9 EUR
APOSOR16996-1G APOSOR16996-5G APOSOR16996-25G
5-Bromo-3-chloropyridine-2-carboxylic acid 97%
5-Bromo-3-chloropyridine-2-carboxylic acid
Formel: C₆H₃BrClNO₂
Molecular Weight: 236.46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827552
CAS-Nummer: 1189513-51-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR