3,5-Dichlor-4-hydroxybenzonitril 97%

Lieferant: Apollo Scientific

Synonyme: Chloroxynil

OR17010-1G OR17010-5G
APOSOR17010-1GEA 62.5 EUR
APOSOR17010-1G APOSOR17010-5G
3,5-Dichlor-4-hydroxybenzonitril 97%
3,5-Dichlor-4-hydroxybenzonitril
Formel: C₇H₃Cl₂NO
Schmelzpunkt: 143…146 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002177
CAS-Nummer: 1891-95-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR