3-Brom-p-kresol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Brom-3-hydroxy-6-methylbenzol, 3-Brom-4-methylphenol, 2-Brom-4-hydroxytoluol
OR17017-1G
OR17017-25G
OR17017-5G
APOSOR17017-1GEA
21.9
EUR
APOSOR17017-1G
APOSOR17017-25G
APOSOR17017-5G
3-Brom-p-kresol 98%
3-Brom-p-kresol
Formel:
C₇H₇BrO Molecular Weight: 187.04 g/mol Schmelzpunkt: 54…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08273793 CAS-Nummer: 60710-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...