3-Brom-p-kresol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Brom-3-hydroxy-6-methylbenzol, 3-Brom-4-methylphenol, 2-Brom-4-hydroxytoluol

OR17017-1G OR17017-25G OR17017-5G
APOSOR17017-1GEA 21.9 EUR
APOSOR17017-1G APOSOR17017-25G APOSOR17017-5G
3-Brom-p-kresol 98%
3-Brom-p-kresol
Formel: C₇H₇BrO
Molecular Weight: 187.04 g/mol
Schmelzpunkt: 54…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08273793
CAS-Nummer: 60710-39-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR