4-Bromoindan-2-one 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1H-inden-2(3H)-one, 4-Bromo-1,3-dihydro-2H-inden-2-one, 4-Bromo-2,3-dihydro-2-oxo-1H-indene, 4-Bromo-2-oxoindane

OR17022-250MG
APOSOR17022-250MGEA 65.4 EUR
APOSOR17022-250MG
4-Bromoindan-2-one 97%
4-Bromoindan-2-one
Formel: C₉H₇BrO
Molecular Weight: 211.055 g/mol
Schmelzpunkt: 74…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09744095
CAS-Nummer: 846032-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR