Ethyl-2-methylthiazol-5-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Ethoxycarbonyl)-2-methyl-1,3-thiazole, Ethyl 2-methylthiazole-5-carboxylate, Ethyl 2-methyl-1,3-thiazole-5-carboxylate

OR17034-500MG
APOSOR17034-500MGEA 21.9 EUR
APOSOR17034-500MG
Ethyl-2-methylthiazol-5-carboxylat
Ethyl-2-methylthiazol-5-carboxylat
Formel: C₇H₉NO₂S
Molecular Weight: 171,2200 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03840442
CAS-Nummer: 79836-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR