2-Chlor-6-methyl-4-pyridinboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

OR17039-1G
APOSOR17039-1GEA 275 EUR
APOSOR17039-1G
2-Chlor-6-methyl-4-pyridinboronsäurepinakolester 98%
2-Chlor-6-methyl-4-pyridinboronsäurepinakolester
Formel: C₁₂H₁₇BClNO₂
Molecular Weight: 253,5329 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08063106
CAS-Nummer: 697739-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR