2-Hydroxy-7-methoxyquinoline-3-carboxaldehyde 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxy-7-methoxyquinoline-3-carbaldehyde, 3-Formyl-2-hydroxy-7-methoxyquinoline, 3-Formyl-2-hydroxy-7-methoxy-1-azanaphthalene

OR17045-250MG
APOSOR17045-250MGEA 150 EUR
APOSOR17045-250MG
2-Hydroxy-7-methoxyquinoline-3-carboxaldehyde 95+%
2-Hydroxy-7-methoxyquinoline-3-carboxaldehyde
Formel: C₁₁H₉NO₃
Molecular Weight: 203,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02986395
CAS-Nummer: 101382-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR