5-Pyridin-3-yl-1H-pyrazole-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR17049-1G
APOSOR17049-1GEA
89.9
EUR
APOSOR17049-1G
5-Pyridin-3-yl-1H-pyrazole-3-carboxylic acid
5-Pyridin-3-yl-1H-pyrazole-3-carboxylic acid
Formel:
C₉H₇N₃O₂ Schmelzpunkt: 308…310 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05170031 CAS-Nummer: 374064-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...