2-Methylsulphanyl-7H-pyrrolo[2,3-d]pyrimidin-4-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydroxy-2-(methylthio)-7H-pyrrolo[2,3-d]pyrimidine

OR17056-500MG
APOSOR17056-500MGEA 21.9 EUR
APOSOR17056-500MG
2-Methylsulphanyl-7H-pyrrolo[2,3-d]pyrimidin-4-ol
2-Methylsulphanyl-7H-pyrrolo[2,3-d]pyrimidin-4-ol
Formel: C₇H₇N₃OS
Molecular Weight: 181,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02668244
CAS-Nummer: 67831-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR