4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-chloro-7H-pyrrolo[2,3-d]pyrimidine

OR17061-1G
APOSOR17061-1GEA 21.9 EUR
APOSOR17061-1G
4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine 95+%
4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine
Formel: C₆H₅ClN₄
Molecular Weight: 168,58 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07369229
CAS-Nummer: 84955-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR