Ethyl-5-nitro-1H-pyrrole-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-nitro-1H-pyrrole-2-carboxylate

OR17066-500MG
APOSOR17066-500MGEA 424 EUR
APOSOR17066-500MG
Ethyl-5-nitro-1H-pyrrole-2-carboxylate
Ethyl-5-nitro-1H-pyrrole-2-carboxylate
Formel: C₇H₈N₂O₄
Molecular Weight: 184,15 g/mol
Schmelzpunkt: 146 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025771
CAS-Nummer: 36131-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR