2-Nitro-4-pyridinecarboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Nitroisonicotinic acid
OR17067-1G
APOSOR17067-1GEA
21.9
EUR
APOSOR17067-1G
2-Nitro-4-pyridinecarboxylic acid
2-Nitro-4-pyridinecarboxylic acid
Formel:
C₆H₄N₂O₄ Molecular Weight: 168,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114160 CAS-Nummer: 33225-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...