1-Boc-7-benzyloxy-3-hydroxymethylindole 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-(Benzyloxy)-3-(hydroxymethyl)-1H-indole, N-BOC protected

OR1707-1G
APOSOR1707-1GEA 232 EUR
APOSOR1707-1G
1-Boc-7-benzyloxy-3-hydroxymethylindole 98%
1-Boc-7-benzyloxy-3-hydroxymethylindole
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353,42 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864726
CAS-Nummer: 914349-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR