Quinoxaline-5-carboxaldehyde 95+%

Lieferant: Apollo Scientific

Synonyme: Quinoxaline-5-carbaldehyde, 5-Formylquinoxaline

OR17071-250MG
APOSOR17071-250MGEA 71.2 EUR
APOSOR17071-250MG
Quinoxaline-5-carboxaldehyde 95+%
Quinoxaline-5-carboxaldehyde
Formel: C₉H₆N₂O
Molecular Weight: 158,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00805833
CAS-Nummer: 141234-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR