5-Bromo-3-iodo-1H-indole, N-BOC protected 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1-(tert-butoxycarbonyl)-3-iodo-1H-indole, tert-Butyl 5-bromo-3-iodo-1H-indole-1-carboxylate

OR1711-500MG
APOSOR1711-500MGEA 21.9 EUR
APOSOR1711-500MG
5-Bromo-3-iodo-1H-indole, N-BOC protected 97%
5-Bromo-3-iodo-1H-indole, N-BOC protected
Formel: C₁₃H₁₃BrINO₂
Molecular Weight: 422.056 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05864761
CAS-Nummer: 850349-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR