Methyl 3-bromo-5-iodobenzoate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR17113-1G
APOSOR17113-1GEA
21.9
EUR
APOSOR17113-1G
Methyl 3-bromo-5-iodobenzoate 97%
Methyl 3-bromo-5-iodobenzoate
Formel:
C₈H₆BrIO₂ Molecular Weight: 340,941 g/mol Schmelzpunkt: 59…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00673016 CAS-Nummer: 188813-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...