Methyl-3-brom-5-methoxybenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-bromo-5-methoxybenzoate
OR17115-1G
APOSOR17115-1GEA
142
EUR
APOSOR17115-1G
Methyl-3-brom-5-methoxybenzoat
Methyl-3-brom-5-methoxybenzoat
Formel:
C₉H₉BrO₃ Siedepunkt: 156…157 °C/4mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD11042779 CAS-Nummer: 56709-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...