5-Acetamido-2-hydroxybenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Acetylamino)-2-hydroxybenzoic acid, N-Acetyl mesalazine, 5-Acetamidosalicylic acid

OR17121-1G
APOSOR17121-1GEA 0 EUR
APOSOR17121-1G
5-Acetamido-2-hydroxybenzoic acid
5-Acetamido-2-hydroxybenzoic acid
Formel: C₉H₉NO₄
Lagertemperatur: Kühlschrank
MDL: MFCD00463101
CAS-Nummer: 51-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR