3-Iodo-5-methyl-1H-indole, N-BOC protected 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(tert-Butoxycarbonyl)-3-iodo-5-methyl-1H-indole, tert-Butyl 3-iodo-5-methyl-1H-indole-1-carboxylate

OR1715-1G
APOSOR1715-1GEA 209 EUR
APOSOR1715-1G
3-Iodo-5-methyl-1H-indole, N-BOC protected 98%
3-Iodo-5-methyl-1H-indole, N-BOC protected
Formel: C₁₄H₁₆INO₂
Molecular Weight: 357,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864765
CAS-Nummer: 914349-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR