4-Chloro-2-iodo-6-methylphenol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-2-hydroxy-3-iodotoluene, 4-Chloro-6-iodo-o-cresol

OR17198-1G OR17198-5G
APOSOR17198-1GEA 415 EUR
APOSOR17198-1G APOSOR17198-5G
4-Chloro-2-iodo-6-methylphenol 98%
4-Chloro-2-iodo-6-methylphenol
Formel: C₇H₆ClIO
Molecular Weight: 268.479 g/mol
Schmelzpunkt: 59…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16606080
CAS-Nummer: 1257535-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR