5-Bromoquinoxaline 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR17203-1G
OR17203-250MG
OR17203-5G
APOSOR17203-1GEA
116
EUR
APOSOR17203-1G
APOSOR17203-250MG
APOSOR17203-5G
5-Bromoquinoxaline 95%
5-Bromoquinoxaline
Formel:
C₈H₅BrN₂ Schmelzpunkt: 66…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02181615 CAS-Nummer: 76982-23-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...