2-Aminopyridine-4-boronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

OR17204-1G OR17204-100MG OR17204-5G
APOSOR17204-1GEA 241 EUR
APOSOR17204-1G APOSOR17204-100MG APOSOR17204-5G
2-Aminopyridine-4-boronic acid pinacol ester
2-Aminopyridine-4-boronic acid pinacol ester
Formel: C₁₁H₁₇BN₂O₂
Schmelzpunkt: 161…162 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD09607735
CAS-Nummer: 1195995-72-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR