1-Methyl-1H-pyrazol-5-boronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

OR17209-1G
APOSOR17209-1GEA 0 EUR
APOSOR17209-1G
1-Methyl-1H-pyrazol-5-boronsäurepinakolester
1-Methyl-1H-pyrazol-5-boronsäurepinakolester
Formel: C₁₀H₁₇BN₂O₂
Schmelzpunkt: 67…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05861380
CAS-Nummer: 847818-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR